Gene Transcripts

Gene Symbol
Triml2
Entrez Gene ID
622117
Gene Name
tripartite motif family-like 2
Gene Aliases
EG622117, Tu4
Chromosome Location
Chr. 8: 43180506 - 43193884 on Build GRCm38
UniGene ID
Mm.390031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160412.1 NP_001153884.1 1 165
GenBank mRNA AV461889.1 - 1 175
EU599038.1 - 3 255
EU599039.1 - 2 239
EU599040.1 - 2 239

View Full Product Details