Gene Transcripts

Gene Symbol
Vmn2r18
Entrez Gene ID
632671
Gene Name
vomeronasal 2, receptor 18
Gene Aliases
EG632671, Vn2r18
Chromosome Location
Chr. 5: 151561661 - 151586924 on Build GRCm38
UniGene ID
Mm.469837
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102582.1 NP_001096052.1 2 579

View Full Product Details