Gene Transcripts

Gene Symbol
Kncn
Entrez Gene ID
654462
Gene Name
kinocilin
Gene Aliases
Kino, L5
Chromosome Location
Chr. 4: 115884400 - 115887964 on Build GRCm38
UniGene ID
Mm.388913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039124.3 NP_001034213.1 2, 3 546
GenBank mRNA BC132034.1 - 2, 3 493
BC132036.1 - 2, 3 493 40130313
DQ235452.1 - 2, 3 546

View Full Product Details