Gene Transcripts

Gene Symbol
Angptl7
Entrez Gene ID
654812
Gene Name
angiopoietin-like 7
Gene Aliases
Chromosome Location
Chr. 4: 148492979 - 148500462 on Build GRCm38
UniGene ID
Mm.388929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039554.3 NP_001034643.1 1, 2 520
GenBank mRNA AK031675.1 - 1, 2 520
BC023373.1 - 1, 2 507 5043159

View Full Product Details