Gene Transcripts

Gene Symbol
Hspbp1
Entrez Gene ID
66245
Gene Name
HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1
Gene Aliases
1500019G21Rik
Chromosome Location
Chr. 7: 4660515 - 4685121 on Build GRCm38
UniGene ID
Mm.6803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024172.3 NP_077134.1 5 839
GenBank mRNA AF338351.1 - 4 777
AK013263.1 - 5 830
AK088469.1 - 5 815
AK160128.1 - 5 834
BC014758.1 - 5 839 4014661

View Full Product Details