Gene Transcripts

Gene Symbol
Ccdc28b
Entrez Gene ID
66264
Gene Name
coiled coil domain containing 28B
Gene Aliases
1110002E08Rik, 1810010N17Rik
Chromosome Location
Chr. 4: 129619274 - 129623908 on Build GRCm38
UniGene ID
Mm.383215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025455.2 NP_079731.2 4, 5 641
GenBank mRNA AK007427.1 - 3, 4 418
AK009176.1 - 2, 3 261
AK028262.1 - 4, 5 619
AK158016.1 - 4, 5 645
BC028873.1 - 4, 5 633 4220355
BQ031123.1 - 2, 3 212

View Full Product Details