Gene Transcripts
- Gene Symbol
- Vmn2r5
- Entrez Gene ID
- 667060
- Gene Name
- vomeronasal 2, receptor 5
- Gene Aliases
- EG667060
- Chromosome Location
- Chr. 3: 64490783 - 64534962 on Build GRCm38
- UniGene ID
- Mm.484957
- Species
- Mus musculus
- Gene Symbol
- Vmn2r-ps11
- Entrez Gene ID
- 319210
- Gene Name
- vomeronasal 2, receptor, pseudogene 11
- Gene Aliases
- 4930518C23Rik
- Chromosome Location
- Chr. 3: 64632863 - 64844743 on Build GRCm38
- UniGene ID
- Mm.246646
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029734.1 |
- |
4 |
543 |
|
BB615497.1 |
- |
4 |
543 |
|
View Full Product Details