Gene Transcripts

Gene Symbol
Pex10
Entrez Gene ID
668173
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
AV128229, Gm142
Chromosome Location
Chr. 4: 155067030 - 155072406 on Build GRCm38
UniGene ID
Mm.133114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042407.1 NP_001035866.1 4, 5 810
GenBank mRNA BC147304.1 - 4, 5 795
BC147305.1 - 4, 5 795 9056374
BY385428.1 - 1, 2 170
CB203398.1 - 3, 4 384

View Full Product Details