Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001097621.1 | NP_001091090.1 | 13 | 5429 | |
GenBank mRNA | AK129315.1 | - | 5 | 3556 | |
BC059062.1 | - | 2 | 2260 | ||
BC094358.1 | - | 2 | 2934 | ||
GQ489027.1 | - | 13 | 5272 |