Gene Transcripts
- Gene Symbol
- Shisa5
- Entrez Gene ID
- 66940
- Gene Name
- shisa family member 5
- Gene Aliases
- 2310008D10Rik, 6430628I05Rik, Scotin, mShisa5
- Chromosome Location
- Chr. 9: 109038567 - 109057792 on Build GRCm38
- UniGene ID
- Mm.196533
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284332.1 |
NP_001271261.1
|
4, 5 |
568 |
|
NM_025858.3 |
NP_080134.1
|
4, 5 |
565 |
|
NM_026381.5 |
NP_080657.1
|
2, 3 |
265 |
|
GenBank mRNA |
AF520699.1 |
- |
4, 5 |
547 |
|
AK009225.1 |
- |
4, 5 |
505 |
|
AK018291.1 |
- |
2, 3 |
221 |
|
AK020877.1 |
- |
2, 3 |
242 |
|
AK033545.1 |
- |
4, 5 |
525 |
|
AK045368.1 |
- |
4, 5 |
495 |
|
AK081060.1 |
- |
2, 3 |
243 |
|
AK088188.1 |
- |
3, 4 |
261 |
|
AK090357.1 |
- |
2, 3 |
419 |
|
AK090373.1 |
- |
4, 5 |
504 |
|
AK205406.1 |
- |
1, 2 |
137 |
|
AK212074.1 |
- |
2, 3 |
224 |
|
BC010238.1 |
- |
4, 5 |
529 |
3581921 |
BC017600.1 |
- |
2, 3 |
177 |
4487600 |
BC057089.1 |
- |
2, 3 |
217 |
5689110 |
View Full Product Details