Gene Transcripts
- Gene Symbol
- Tctex1d1
- Entrez Gene ID
- 67344
- Gene Name
- Tctex1 domain containing 1
- Gene Aliases
- 1700055O19Rik
- Chromosome Location
- Chr. 4: 102978388 - 103005594 on Build GRCm38
- UniGene ID
- Mm.111346
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163767.1 |
NP_001157239.1
|
3, 4 |
415 |
|
NM_001163768.1 |
NP_001157240.1
|
2, 3 |
322 |
|
NM_026100.3 |
NP_080376.1
|
3, 4 |
447 |
|
GenBank mRNA |
AK006804.1 |
- |
3, 4 |
416 |
|
AK015315.1 |
- |
3, 4 |
450 |
|
BB634690.1 |
- |
2, 3 |
334 |
|
BC048635.1 |
- |
3, 4 |
387 |
|
BC099549.1 |
- |
3, 4 |
421 |
30919464 |
BC132398.1 |
- |
3, 4 |
365 |
40130675 |
BC132400.1 |
- |
3, 4 |
365 |
40130677 |
BC145187.1 |
- |
3, 4 |
365 |
9053738 |
View Full Product Details