Gene Transcripts

Gene Symbol
Nmral1
Entrez Gene ID
67824
Gene Name
NmrA-like family domain containing 1
Gene Aliases
1110025F24Rik, AI256624
Chromosome Location
Chr. 16: 4711317 - 4719356 on Build GRCm38
UniGene ID
Mm.28451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026393.2 NP_080669.1 3 713
GenBank mRNA BC030039.1 - 3 416

View Full Product Details