Gene Transcripts

Gene Symbol
Slc25a39
Entrez Gene ID
68066
Gene Name
solute carrier family 25, member 39
Gene Aliases
3010027G13Rik, D11Ertd333e
Chromosome Location
Chr. 11: 102402969 - 102408675 on Build GRCm38
UniGene ID
Mm.44226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026542.3 NP_080818.1 12 1144
GenBank mRNA AK002381.1 - 12 1099
AK007276.1 - 12 1122
AK007934.1 - 12 1098
AK010272.1 - 12 1120
AK013834.1 - 8 788
AK019396.1 - 12 1136
AK078417.1 - 12 1136
AK084116.1 - 11 1212
AK160922.1 - 12 1090
AK161289.1 - 12 1136
BC115661.1 - 12 1044 40103267
BC115662.1 - 12 1051 40103268

View Full Product Details