Gene Transcripts

Gene Symbol
Tatdn3
Entrez Gene ID
68972
Gene Name
TatD DNase domain containing 3
Gene Aliases
1500010M24Rik
Chromosome Location
Chr. 1: 191045826 - 191063010 on Build GRCm38
UniGene ID
Mm.237188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163421.1 NP_001156893.1 4 286
NM_001163422.1 NP_001156894.1 4 286
NM_026895.1 NP_081171.1 4 286
GenBank mRNA AK005193.1 - 4 257
AK156071.1 - 4 237
BC061248.1 - 4 258

View Full Product Details