Gene Transcripts

Gene Symbol
Spcs1
Entrez Gene ID
69019
Gene Name
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases
1810004F21Rik
Chromosome Location
Chr. 14: 30999826 - 31001666 on Build GRCm38
UniGene ID
Mm.28852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026911.3 NP_081187.2 3 566
GenBank mRNA AI848281.1 - 2 453
AK007336.1 - 3 264
AK082928.1 - 3 265
AK088658.1 - 2 349
BC020050.1 - 468
BC034270.2 - 3 441
BC049895.1 - 3 257 3991764

View Full Product Details