Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190453.1 | NP_001177382.1 | 1, 2 | 283 | |
NM_001190454.1 | NP_001177383.1 | 1, 2 | 283 | ||
NM_027151.2 | NP_081427.1 | 1, 2 | 283 | ||
GenBank mRNA | AK054390.1 | - | 1, 2 | 177 | |
AK144464.1 | - | 1, 2 | 159 | ||
AK144505.1 | - | 1, 2 | 159 | ||
AK164021.1 | - | 1, 2 | 160 | ||
BC004613.1 | - | 1, 2 | 146 | 3585310 |