Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027210.3 | NP_081486.1 | 3 | 849 | |
NM_028171.2 | NP_082447.1 | 3 | 849 | ||
GenBank mRNA | AK005430.1 | - | 3 | 840 | |
AK005537.1 | - | 3 | 850 | ||
AK146045.1 | - | 3 | 850 | ||
BC099430.1 | - | 3 | 836 | 30787167 |