Gene Transcripts

Gene Symbol
Tmem70
Entrez Gene ID
70397
Gene Name
transmembrane protein 70
Gene Aliases
1110020A09Rik, 2210416J16Rik
Chromosome Location
Chr. 1: 16665191 - 16678275 on Build GRCm38
UniGene ID
Mm.194225
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026392.1 NP_080668.1 3 724
NM_027415.2 NP_081691.2 3 721
GenBank mRNA AK003839.1 - 3 706
AK005075.1 - 3 706
AK008955.1 - 3 690
AK151280.1 - 3 696
AK167069.1 - 3 647
AK169031.1 - 3 654
BC011320.1 - 3 650 3979025

View Full Product Details