Gene Transcripts

Gene Symbol
Mutyh
Entrez Gene ID
70603
Gene Name
mutY homolog (E. coli)
Gene Aliases
5730495A01Rik, Mutyha, Mutyhalpha, Mutyhb, Mutyhbeta, Mutyhc, Myh
Chromosome Location
Chr. 4: 116807682 - 116819439 on Build GRCm38
UniGene ID
Mm.180333
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159581.1 NP_001153053.1 6 481
NM_133250.2 NP_573513.2 5 388
GenBank mRNA AB117938.1 - 4 350
AB117939.1 - 6 465
AK017730.1 - 5 376
AK077546.1 - 5 385
AK156790.1 - 6 471
AY007717.1 - 4 344
BC057942.1 - 5 369 5320744
CX213088.1 - 5 423

View Full Product Details