Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
100504195
Gene Name
MICAL C-terminal like
Gene Aliases
4921517J23Rik
Chromosome Location
Chr. 7: 112368308 - 112413104 on Build GRCm38
UniGene ID
Mm.291129
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027587.2 NP_081863.2 4 1436
GenBank mRNA AB359922.1 - 4 1436
AK014911.1 - 1 871
BC031386.1 - 4 1610 5043827

View Full Product Details