Gene Transcripts

Gene Symbol
Tysnd1
Entrez Gene ID
71767
Gene Name
trypsin domain containing 1
Gene Aliases
1300019N10Rik
Chromosome Location
Chr. 10: 61695497 - 61702773 on Build GRCm38
UniGene ID
Mm.71514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272090.1 NP_001259019.1 1 536
NM_001272091.1 NP_001259020.1 1 536
NM_001272092.1 NP_001259021.1 1 536
NM_027912.1 NP_082188.1 1 536
GenBank mRNA AK005069.1 - 1 537
AK154513.1 - 1 519
BC020273.1 - 1 527
BC119320.1 - 1 514 8734069

View Full Product Details