Gene Transcripts

Gene Symbol
Bbs10
Entrez Gene ID
71769
Gene Name
Bardet-Biedl syndrome 10 (human)
Gene Aliases
1300007O09Rik, AI452285
Chromosome Location
Chr. 10: 111298679 - 111301736 on Build GRCm38
UniGene ID
Mm.45256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027914.1 NP_082190.1 2 1891
GenBank mRNA AK004937.1 - 1 2197

View Full Product Details