Gene Transcripts

Gene Symbol
Tom1l1
Entrez Gene ID
71943
Gene Name
target of myb1-like 1 (chicken)
Gene Aliases
2310045L10Rik, C80573, Srcasm
Chromosome Location
Chr. 11: 90645690 - 90687661 on Build GRCm38
UniGene ID
Mm.167868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028011.2 NP_082287.1 6 592
GenBank mRNA AF395837.1 - 6 556
AJ505007.1 - 6 553
AK033712.1 - 6 594
AK083630.1 - 554

View Full Product Details