Gene Transcripts

Gene Symbol
Plekhf1
Entrez Gene ID
72287
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 1
Gene Aliases
1810013P09Rik, APPD, LAPF, PHAFIN1, ZFYVE15
Chromosome Location
Chr. 7: 38220654 - 38227994 on Build GRCm38
UniGene ID
Mm.333798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024413.2 NP_077724.2 2 257
GenBank mRNA AK143976.1 - 246
AK157990.1 - 2 246
AK170283.1 - 2 256
AK171398.1 - 2 246
AW227287.1 - 2 257

View Full Product Details