Gene Transcripts

Gene Symbol
Cdkn3
Entrez Gene ID
72391
Gene Name
cyclin-dependent kinase inhibitor 3
Gene Aliases
2410006H10Rik, KAP
Chromosome Location
Chr. 14: 46760464 - 46771529 on Build GRCm38
UniGene ID
Mm.272394
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028222.1 NP_082498.1 6 532
GenBank mRNA AK146312.1 - 6 448
BC049694.1 - 6 516 6705916

View Full Product Details