Gene Transcripts

Gene Symbol
Exo5
Entrez Gene ID
73172
Gene Name
exonuclease 5
Gene Aliases
3110037I16Rik, AV297100, Dem1, Exo V, mExo5
Chromosome Location
Chr. 4: 120921202 - 120925005 on Build GRCm38
UniGene ID
Mm.151436
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160043.1 NP_001153515.1 2 1396
NM_028457.2 NP_082733.1 2 1417
GenBank mRNA AK014134.1 - 2 1415
BC023360.1 - 2 1410 5064775

View Full Product Details