Gene Transcripts

Gene Symbol
Dcbld2
Entrez Gene ID
73379
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
1700055P21Rik, AW146002, CLCP1, Esdn
Chromosome Location
Chr. 16: 58408426 - 58469745 on Build GRCm38
UniGene ID
Mm.373589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028523.3 NP_082799.2 7 1054
GenBank mRNA AF387548.1 - 7 1146
AK051889.1 - 5 332
BC066097.1 - 7 1054 6856458

View Full Product Details