Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028633.3 | NP_082909.1 | 1 | 122 | |
NM_207104.1 | NP_996987.1 | 1 | 122 | ||
GenBank mRNA | AK050729.1 | - | 1 | 56 | |
AY029337.1 | - | 1 | 122 | ||
BC025464.1 | - | 1 | 113 | 5351961 |