Gene Transcripts

Gene Symbol
Serpina3a
Entrez Gene ID
74069
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3A
Gene Aliases
Chromosome Location
Chr. 12: 104106704 - 104121897 on Build GRCm38
UniGene ID
Mm.159128
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167705.1 NP_001161177.1 2 595
GenBank mRNA BC063325.1 - 2 785

View Full Product Details