Gene Transcripts

Gene Symbol
Klrg2
Entrez Gene ID
74253
Gene Name
killer cell lectin-like receptor subfamily G, member 2
Gene Aliases
2310020F24Rik
Chromosome Location
Chr. 6: 38621414 - 38637706 on Build GRCm38
UniGene ID
Mm.121859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033171.2 NP_001028343.2 5 1243
GenBank mRNA AK009417.1 - 5 1244
AK145064.1 - 5 1238
AK162933.1 - 6 831
AK169862.1 - 6 787

View Full Product Details