Gene Transcripts

Gene Symbol
Iqcf1
Entrez Gene ID
74267
Gene Name
IQ motif containing F1
Gene Aliases
1700055J15Rik
Chromosome Location
Chr. 9: 106499967 - 106502250 on Build GRCm38
UniGene ID
Mm.116682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146701.1 NP_001140173.1 2 116
NM_028843.1 NP_083119.1 3 221
GenBank mRNA AK006797.1 - 3 197
BC049769.1 - 95 6774620

View Full Product Details