Gene Transcripts

Gene Symbol
Nkx6-3
Entrez Gene ID
74561
Gene Name
NK6 homeobox 3
Gene Aliases
9130417I07Rik, Nkx6.3
Chromosome Location
Chr. 8: 23153271 - 23158948 on Build GRCm38
UniGene ID
Mm.290028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029002.2 NP_083278.1 1 481
GenBank mRNA AK018683.1 - 1 482
AK147163.1 - 1 453
BC140994.1 - 1 352 9055606
DQ093570.1 - 1 167

View Full Product Details