Gene Transcripts

Gene Symbol
Spdyb
Entrez Gene ID
74673
Gene Name
speedy homolog B (Xenopus laevis)
Gene Aliases
4930445A17Rik, 4930451F05Rik, Spdy2, Spdye4
Chromosome Location
Chr. 5: 143216316 - 143226902 on Build GRCm38
UniGene ID
Mm.273303
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029048.3 NP_083324.3 1 136
GenBank mRNA AK015441.1 - 1 137
AY820308.1 - 1 61

View Full Product Details