Gene Transcripts
- Gene Symbol
- Sepn1
- Entrez Gene ID
- 74777
- Gene Name
- selenoprotein N, 1
- Gene Aliases
- 1110019I12Rik, AI414492
- Chromosome Location
- Chr. 4: 134537892 - 134552166 on Build GRCm38
- UniGene ID
- Mm.29773
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029100.2 |
NP_083376.2
|
6, 7 |
956 |
|
GenBank mRNA |
CX562316.1 |
- |
5, 6 |
633 |
|
View Full Product Details