Gene Transcripts

Gene Symbol
Hspb9
Entrez Gene ID
75482
Gene Name
heat shock protein, alpha-crystallin-related, B9
Gene Aliases
1700007H20Rik
Chromosome Location
Chr. 11: 100713850 - 100714575 on Build GRCm38
UniGene ID
Mm.46175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029307.1 NP_083583.1 1 562
GenBank mRNA AK005714.1 - 1 485

View Full Product Details