Gene Transcripts

Gene Symbol
Morn5
Entrez Gene ID
75495
Gene Name
MORN repeat containing 5
Gene Aliases
1700010A17Rik
Chromosome Location
Chr. 2: 36049473 - 36079709 on Build GRCm38
UniGene ID
Mm.276332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029309.2 NP_083585.1 4 455
GenBank mRNA AK005808.1 - 4 443
BC036965.1 - 4 461 4511210
BC048673.1 - 4 455 6774106

View Full Product Details