Gene Transcripts

Gene Symbol
Tm4sf5
Entrez Gene ID
75604
Gene Name
transmembrane 4 superfamily member 5
Gene Aliases
2010003F10Rik, AI323987
Chromosome Location
Chr. 11: 70505274 - 70511183 on Build GRCm38
UniGene ID
Mm.24400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029360.3 NP_083636.2 4 586
GenBank mRNA AI323987.1 - 2 393
AK008064.1 - 4 584
BC010782.1 - 4 546 4219239

View Full Product Details