Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001048060.2 | NP_001041525.1 | 2, 3 | 122 | |
NM_198004.3 | NP_932121.2 | 2, 3 | 122 | ||
GenBank mRNA | AK017224.1 | - | 2, 3 | 117 | |
AK032811.1 | - | 2, 3 | 105 | ||
AK040588.1 | - | 2, 3 | 123 | ||
AK045763.1 | - | 2, 3 | 118 | ||
BC026742.1 | - | 2, 3 | 91 | 4192995 | |
BU525297.1 | - | 2, 3 | 125 | ||
CX206109.1 | - | 2, 3 | 98 |