Gene Transcripts

Gene Symbol
Kdm8
Entrez Gene ID
77035
Gene Name
lysine (K)-specific demethylase 8
Gene Aliases
3110005O21Rik, Jmjd5
Chromosome Location
Chr. 7: 125444620 - 125462269 on Build GRCm38
UniGene ID
Mm.307239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029842.5 NP_084118.1 5 986
GenBank mRNA AK014006.1 - 5 930
BC024807.1 - 5 914 5355270

View Full Product Details