Gene Transcripts

Gene Symbol
Notum
Entrez Gene ID
77583
Gene Name
notum pectinacetylesterase homolog (Drosophila)
Gene Aliases
5730593N15Rik
Chromosome Location
Chr. 11: 120653788 - 120661745 on Build GRCm38
UniGene ID
Mm.32839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175263.4 NP_780472.3 4 504
GenBank mRNA AK019987.1 - 4 504
DN174397.1 - 4 510

View Full Product Details