Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029975.2 | NP_084251.1 | 3 | 522 | |
GenBank mRNA | AK020784.1 | - | 3 | 397 | |
AK037408.1 | - | 3 | 494 | ||
AK084712.1 | - | 621 | |||
AK085119.1 | - | 684 | |||
AY249021.1 | - | 2 | 151 | ||
BC132070.1 | - | 3 | 522 | 40130347 | |
BC132072.1 | - | 3 | 522 | 40130349 | |
EU047812.1 | - | 3 | 386 |