Gene Transcripts

Gene Symbol
Tnfrsf23
Entrez Gene ID
79201
Gene Name
tumor necrosis factor receptor superfamily, member 23
Gene Aliases
SOB, TNFRSFH23, Tnfrh1, Tnfrsf1al1, mDcTRAILR1, mSOB
Chromosome Location
Chr. 7: 143665807 - 143686423 on Build GRCm38
UniGene ID
Mm.290780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ278264.1 - 1, 2 61
AK154786.1 - 1, 2 87
AK170037.1 - 1, 2 90
AY046550.1 - 1, 2 70
AY165625.1 - 1, 2 76
BC106828.1 - 1, 2 72 40044453
BC106829.1 - 1, 2 76 40044455
BG077775.2 - 1, 2 62
BY036522.1 - 1, 2 86
BY201915.1 - 1, 2 92

View Full Product Details