Gene Transcripts

Gene Symbol
Hist1h1a
Entrez Gene ID
80838
Gene Name
histone cluster 1, H1a
Gene Aliases
H1.1, H1a, H1f1, H1var3
Chromosome Location
Chr. 13: 23763668 - 23764412 on Build GRCm38
UniGene ID
Mm.480242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030609.3 NP_085112.1 1 74
GenBank mRNA BC116820.1 - 1 49
BC116850.1 - 1 47 40126169

View Full Product Details