Gene Transcripts

Gene Symbol
Abo
Entrez Gene ID
80908
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase)
Gene Aliases
NAGAT
Chromosome Location
Chr. 2: 26842496 - 26864995 on Build GRCm38
UniGene ID
Mm.160386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030718.5 NP_109643.3 5 407
GenBank mRNA AB041039.1 - 4 295
AK033786.1 - 5 328
AK035261.1 - 5 392
BC116759.1 - 5 356 40126078
BC116761.1 - 5 356 40126080

View Full Product Details