Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024281.2 | NP_077243.2 | 2 | 2093 | |
NM_133626.2 | NP_598329.1 | 2 | 2093 | ||
GenBank mRNA | AF273683.1 | - | 2439 | ||
AF273684.1 | - | 2 | 2016 | ||
AF273685.1 | - | 3 | 1836 | ||
AF273686.1 | - | 3 | 1053 | ||
AF273687.1 | - | 3 | 1053 | ||
AF273688.1 | - | 4 | 1107 | ||
AF273689.1 | - | 3 | 903 | ||
AK019964.1 | - | 1 | 26 | ||
BC031452.1 | - | 2 | 2082 |