Gene Transcripts

Gene Symbol
Tex16
Entrez Gene ID
83556
Gene Name
testis expressed gene 16
Gene Aliases
OTTMUSG00000018437
Chromosome Location
Chr. X: 111894858 - 112127326 on Build GRCm38
UniGene ID
Mm.190608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031382.2 NP_113559.2 3 1879
GenBank mRNA AF285573.1 - 3 1879
BC132332.1 - 3 1903 40130609

View Full Product Details