Gene Transcripts

Gene Symbol
Pcdhb13
Entrez Gene ID
93884
Gene Name
protocadherin beta 13
Gene Aliases
Pcdbh6, PcdhbM
Chromosome Location
Chr. 18: 37442517 - 37446209 on Build GRCm38
UniGene ID
Mm.196700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053138.2 NP_444368.1 1 414
GenBank mRNA AK082435.1 - 1 416
AY013774.1 - 1 360
BC138123.1 - 1 431 8861142
BC138124.1 - 1 431 8861143

View Full Product Details