Gene Transcripts
- Gene Symbol
- Eps8l3
- Entrez Gene ID
- 99662
- Gene Name
- EPS8-like 3
- Gene Aliases
- AI504489, Eps8r3
- Chromosome Location
- Chr. 3: 107873860 - 107892900 on Build GRCm38
- UniGene ID
- Mm.108491
- Species
- Mus musculus
- Gene Symbol
- 4933431E20Rik
- Entrez Gene ID
- 329735
- Gene Name
- RIKEN cDNA 4933431E20 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 107888850 - 107896213 on Build GRCm38
- UniGene ID
- Mm.270500
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
NM_177858.1 |
- |
|
2078 |
|
Chr. - - [-] |
View Full Product Details