Product Details

SNP ID
rs13481701
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:14521082 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGAATAAATTTAGGCAACTGTCTTC[A/G]TGAAAATCTGTCCCCCCACAAAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Hecw1 PubMed Links

Gene Details

Gene
Hecw1
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081348.3 Intron NP_001074817.3

View Full Product Details