Product Details

SNP ID
rs4223977
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:57458578 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCCTATCAGAATTCACTGCCTGAGG[C/T]CAGCTTTGTATGGCTTAGAATCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Wwtr1 PubMed Links

Gene Details

Gene
Wwtr1
Gene Name
WW domain containing transcription regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168281.1 1816 UTR 3 NP_001161753.1
NM_133784.3 1816 UTR 3 NP_598545.2

View Full Product Details